Pyridinium tribromide ≥85.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

P0825-500G P0825-25G
TCP0825-500GEA 322.36 USD
TCP0825-500G TCP0825-25G
Pyridinium tribromide ≥85.0% (by titrimetric analysis)
Pyridinium tribromide

CAS Number: 39416-48-3
MDL Number: MFCD00013223
Molecular Formula: C5H6Br3N
Molecular Weight: 319.82
Purity/Analysis Method: >85.0% (T)
Form: Crystal
Melting point (°C): 134


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Pyridine Hydrobromide Perbromide, Pyridinium Tribromide

Formula: C₅H₆Br₃N
MW: 319.82 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00013223
CAS Number: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34599efa161a53786b27ab6e6d85ee6a381f857b&vid=34599efa161a53786b27ab6e6d85ee6a381f857b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR