Propargyl amine ≥97.0%

Supplier: TCI America

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

P0911-5ML P0911-25ML
TCP0911-005MLEA 63.4 USD
TCP0911-005ML TCP0911-025ML
Propargyl amine ≥97.0%
Propargyl amine

CAS Number: 2450-71-7
MDL Number: MFCD00008198
Molecular Formula: C3H5N
Molecular Weight: 55.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 84
Flash Point (°C): 6
Specific Gravity (20/20): 0.86
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction), Terminal Alkynes (Click Chemistry), Bioscience, Crosslinkers
Other Product Names / Synonyms: 3-Amino-1-propyne

Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 83
Density: 0.86 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00008198
CAS Number: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1efbedb74cd0e26464a7e8c2adb22ee46774a940&vid=1efbedb74cd0e26464a7e8c2adb22ee46774a940&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR