Pyridinium chlorochromate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
PCC
P0930-500G
P0930-100G
P0930-25G
TCP0930-500GEA
151.7
USD
TCP0930-500G
TCP0930-100G
TCP0930-25G
Pyridinium chlorochromate ≥98.0% (by titrimetric analysis)
Pyridinium chlorochromate
CAS Number: 26299-14-9
MDL Number: MFCD00013106
Molecular Formula: C5H6ClCrNO3
Molecular Weight: 215.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Slightly Pale Yellow Red
Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: PCC
Formula:
C₅H₆ClCrNO₃ MW: 215.56 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD00013106 CAS Number: 26299-14-9 EINECS: 247-595-5 UN: 1479 ADR: 5.1,II Merck Index: 13,08064 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...