3-Propoxypicolinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P1195-1G
P1195-100MG
TCP1195-1GEA
164
USD
TCP1195-1G
TCP1195-100MG
3-Propoxypicolinic acid ≥98.0% (by titrimetric analysis)
3-Propoxypicolinic acid
CAS Number: 14440-94-9
MDL Number: MFCD00191574
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 118
Other Product Names / Synonyms: 3-Propoxypicolinic Acid
Formula:
C₉H₁₁NO₃ MW: 181.188 g/mol Melting Pt: 118 °C |
MDL Number:
MFCD00191574 CAS Number: 14440-94-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...