Tetrabutylammonium azide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

T0920-5G T0920-25G
TCT0920-005GEA 96 USD
TCT0920-005G TCT0920-025G
Tetrabutylammonium azide ≥95.0% (by titrimetric analysis)
Tetrabutylammonium azide

CAS Number: 993-22-6
MDL Number: MFCD00060069
Molecular Formula: C16H36N4
Molecular Weight: 284.49
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 85
Storage Temperature: 0-10°C

Formula: C₁₆H₃₆N₄
Melting Pt: 85 °C
MDL Number: MFCD00060069
CAS Number: 993-22-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f7d23ec3f90efbd42943c1511666d8ec0e984e&vid=53f7d23ec3f90efbd42943c1511666d8ec0e984e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR