1,2,3,4-Tetrachlorobenzene ≥96.0%

Supplier: TCI America
Warning

T0631-500G T0631-25G
TCT0631-500GEA 303.4 USD
TCT0631-500G TCT0631-025G
1,2,3,4-Tetrachlorobenzene ≥96.0%
1,2,3,4-Tetrachlorobenzene

CAS Number: 634-66-2
MDL Number: MFCD00000538
Molecular Formula: C6H2Cl4
Molecular Weight: 215.88
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 254
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₆H₂Cl₄
Boiling Pt: 254 °C
Melting Pt: 45 °C
Density: 1.73 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000538
CAS Number: 634-66-2
EINECS: 211-214-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=081430d9530cf3516d10ec875e0d4d4558a69189&vid=969b35f7e2b0c0de351f5f4a509c3cc14d44cae6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR