Triisobutylene (mixture of branched chain isomer) ≥90.0%

Supplier: TCI America

Synonyms: Isobutylene Trimer, Isobutene Trimer

T0456-25ML T0456-500ML
TCT0456-025MLEA 49 USD
TCT0456-025ML TCT0456-500ML
Triisobutylene (mixture of branched chain isomer) ≥90.0%
Triisobutylene (mixture of branched chain isomer)

CAS Number: 7756-94-7
MDL Number: MFCD00060236
Molecular Formula: C12H24
Molecular Weight: 168.32
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 177
Flash Point (°C): 50
Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: Isobutene Trimer, Isobutylene Trimer

Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 177 °C
Density: 0.77 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00060236
CAS Number: 7756-94-7

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR