4-tert-Octylphenol ≥95.0%
Supplier: TCI America
|
Danger
|
CAS Number: 140-66-9
MDL Number: MFCD00002368
Molecular Formula: C14H22O
Molecular Weight: 206.33
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 279
Melting point (°C): 83
Flash Point (°C): 145
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials, Analytical Science, Alkylphenols (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: 4-tert-Octylphenol
Formula:
C₁₄H₂₂O Boiling Pt: 279 °C Melting Pt: 83 °C Density: 0.932 g/cm³ (20 °C) Flash Pt: 145 °C |
MDL Number:
MFCD00002368 CAS Number: 140-66-9 EINECS: 205-426-2 UN: 2430 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...