Tetradecylamine ≥96.0%

Supplier: TCI America
Warning

Synonyms: 1-Tetradecylamine, 1-Aminotetradecane, n-Tetradecylamine, Tetradecan-1-amine

T0090-25G T0090-400G
TCT0090-025GEA 45.7 USD
TCT0090-025G TCT0090-400G
Tetradecylamine ≥96.0%
Tetradecylamine

CAS Number: 2016-42-4
MDL Number: MFCD00008157
Molecular Formula: C14H31N
Molecular Weight: 213.41
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 291
Flash Point (°C): 131
Freezing point (°C): 37


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Myristylamine, 1-Aminotetradecane

Formula: H₃C(CH₂)₁₂CH₂NH₂
MW: 213.41 g/mol
Boiling Pt: 291 °C
Melting Pt: 40 °C
Density: 0.8571 g/cm³ (20 °C)
Flash Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00008157
CAS Number: 2016-42-4
EINECS: 217-950-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e4013a4ac46ad02f5052e5f15b256011fa6e4c5&vid=fe3a98ac7b1b9de7cc300f02b005fe35ef8ffdf4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR