Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, white crystalline powder, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

02103133.1 02103133.2 02103133.5 0210313301 0210313305 0210313310
IC103133.1EA 47.15 USD
IC103133.1 IC103133.2 IC103133.5 IC10313301 IC10313305 IC10313310
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, white crystalline powder, Ultrapure
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Tris have been useful as buffers in a wide variety of biological systems. It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes). It does not precipitate calcium salts and is of value in maintaining solubility of manganese salts. It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid. It is RNAse and DNAse-free. Tris is relatively non-hygroscopic ; but, if needed, it can be dried at 100 °C for up to 4 hours to remove any water.
Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions.


  • Ultra Pure Grade
  • Presentation: White Crystalline Powder
  • Vapor Pressure: Low at ambient temperatures
  • pKa: 8.1 at 25 °C (Lit.)
  • Storage temperature: Store at Room Temperature, desiccate.


Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions

Formula: NH₂C(CH₂OH)₃
Boiling Pt: 219…220 °C (10 mmHg)) (lit.)
Melting Pt: 165…175 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.9%
FTIR Conforms to Standard
Melting Point 165-175°C
pH (5% aq soln) 10-11.5
OD260 nm (5% aq soln) ≤0.1
Loss on Drying ≤1%
Solubility (40% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR