D-(+)-Trehalose dihydrate ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

0210309705 0210309725 0210309791 0210309710
IC10309705EA 49.82 USD
IC10309705 IC10309725 75875-600 76176-986
D-(+)-Trehalose dihydrate ≥98%, white powder
D-(+)-Trehalose dihydrate

D-(+)-Trehalose dihydrate is a natural alpha-linked non –reducing disaccharide formed by an α,α-1,1-glucoside bond between two α-glucose units which yields 2 moles of D-glucose upon acid hydrolysis. Has 90% of the calorific content of sucrose.


  • Protein stabilizing agent


Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro. Oral administration of trehalose has been shown to effectively extend lifespan in a murine Huntington disease model. Also known to accumulate in osmotically-stressed cultured bacterial cells and protect against the adverse effects of dehydration.


Trehalose has also been used in several biopharmaceutical monoclonal antibody formulations. It is also used as a protein-stabilizing agent in research. Use as a cryoprotectant in a variety of cell freezing media.


Soluble in water (50 mg/mL - clear, colourless solution) or hot alcohol.

Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Melting Pt: 203 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0ce5da7710ce2682bcd20f8c01b8320e1ec6ad3&vid=e0ce5da7710ce2682bcd20f8c01b8320e1ec6ad3&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a2ef2d386c7dd63b8c380c420059ea6becabd96&vid=1a2ef2d386c7dd63b8c380c420059ea6becabd96&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Loss on Drying 8.5 - 10.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR