Sebacoyl dichloride ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Sebacoyl chloride

S0030-500ML S0030-25ML
TCS0030-500MLEA 295 USD
TCS0030-500ML TCS0030-25ML
Sebacoyl dichloride ≥95.0% (by GC, titration analysis)
Sebacoyl dichloride

CAS Number: 111-19-3
MDL Number: MFCD00000770
Molecular Formula: C10H16Cl2O2
Molecular Weight: 239.14
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 167
Melting point (°C): -3
Flash Point (°C): 164
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Decanedioyl Chloride, Sebacic Acid Dichloride

Formula: C₁₀H₁₆Cl₂O₂
MW: 239.14 g/mol
Boiling Pt: 167 °C
Melting Pt: –3 °C
Density: 1.12 (20 °C)
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0e0bf5b08fc9d6106953f7b191de31c4c8d2fad&vid=c0e0bf5b08fc9d6106953f7b191de31c4c8d2fad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR