Potassium permanganate 0.0995-0.1005 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
|
|
Synonyms:
Permanganic acid potassium salt
Management of Change (MOC) category = NI
5651-03
5651-02
JT5651-3CS
532.17
USD
JT5651-3
JT5651-2
Potassium permanganate 0.0995-0.1005 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Potassium permanganate
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.
Formula:
KMnO₄ MW: 158.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 3082 ADR: 9 III Merck Index: 12,07824 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 0.0995 - 0.1005 |
Chloride and Chlorate (as Cl) | ≤10 ppm |
Sulfate (SO₄) | ≤20 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
SRM No… |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...