Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

Synonyms: Permanganic acid potassium salt

Management of Change (MOC) category = N

3227-05 3227-01
JT3227-5CS 1781.26 USD
JT3227-5 JT3227-1
Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®
Potassium permanganate

For mercury determination. Lot analysis on label.

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: ? 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KMnO₄) ≥99.0 %
ACS - Insoluble Matter ≤0.2 %
ACS - Chloride and Chlorate (as Cl) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.02 %
Trace Impurities - Mercury (Hg) ≤0.050 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR