2,4-Dichlorophenylhydrazine

Supplier: Matrix Scientific

Synonyms: (2,4-Dichlorophenyl)hydrazine

039894-25G 039894-100G
103601-234EA 415.38 USD
103601-234 103601-332
2,4-Dichlorophenylhydrazine
2,4-Dichlorophenylhydrazine
Formula: C₆H₆Cl₂N₂
MDL Number: MFCD00044514
CAS Number: 13123-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR