Sodium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIALS US
|
Danger
|
Synonyms:
Sodium metaperiodate
Management of Change (MOC) category = N
3756-04
3756-01
JT3756-4CS
6381.63
USD
JT3756-4
JT3756-1
Sodium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS
Sodium periodate
Lot analysis on label.
- Lot analysis on label
Formula:
NaIO₄ MW: 213.89 g/mol Boiling Pt: 300 Melting Pt: 175 Density: 3.87 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 EINECS: 232-197-6 UN: 1479 ADR: 5.1 II Merck Index: 13,08714 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (NaIO₄) (by iodometry) | 99.8 - 100.3 % |
Other Halogens (as Cl) | ≤0.02 % |
Trace Impurities - Manganese (Mn) | ≤3.000 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...