Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (mFluor™ UV375)

Supplier: AAT BIOQUEST INC
100210X0 100210X1
77023-776EA 244.24 USD
77023-776 77024-810
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (mFluor™ UV375)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR