Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ UV375)

Supplier: AAT BIOQUEST INC
104410X0 104410X1
77021-616EA 244.24 USD
77021-616 77024-300
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ UV375)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ UV375
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR