Anti-CD14 Mouse Monoclonal Antibody (mFluor™ UV375) [clone: 61D3]

Supplier: AAT BIOQUEST INC
101410X0 101410X1
77024-334EA 244.24 USD
77024-334 77023-762
Anti-CD14 Mouse Monoclonal Antibody (mFluor™ UV375) [clone: 61D3]
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR