AlphaTec® 15-554 Chemical resistant gloves, Ansell
Supplier: Ansell Healthcare
214308 214306
47747-396CS 744.08 USD
47747-396 32890-009
AlphaTec® 15-554 Chemical resistant gloves, Ansell
Gloves
Type 15-554 gloves are nearly inert to strong solvents including aromatics, aliphatic substances, and chlorinated solvents.
  • Hand-specific
  • Gauntlet cuffs
  • Good resistance to abrasions, and suitable for handling solvents and working with epoxies
  • Smooth finish
  • Liner material: interlock cotton
Gloves are suitable for electronics applications, handling strong organic solvents, and working with epoxies.
Use in the chemical, electronics, and printing industries. Uses include chemical processing and preparation; electronic manufacture; lab work and analyses, especially when resistance to acids is required; and printing.
Gloves also offer good resistance to snags, punctures, and abrasion. Comfortable, knit-lined gloves are lighter than comparable neoprene-coated gloves. Pre-curved fingers and winged thumb.
Certificaciones: ANSI level 1 cut protection. ANSI level 4 abrasion protection.
Envase: Gloves are packaged 1 pair in a bag; 12 bags in a carton.