Strontium carbonate ≥99.994% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
Powder, 22 mesh. Purity based on metal contaminates only.
Purity: 99.994%
CAS No.: 1633-05-2
Molecular Formula: SrCO3
Formula Weight: 147.63
Physical Form: -22 Mesh Powder
Density: 3.70
Melting Point: 1100° dec.
Merck Reference: 14,8838
MDL No.: MFCD00011250
Formula:
CO₃Sr Boiling Pt: 2647 °C (1 mmHg) Melting Pt: 1100 °C (dec.) Density: 3.70 Storage Temperature: Ambient |
MDL Number:
MFCD00011250 CAS Number: 1633-05-2 EINECS: 216-643-7 Merck Index: 14,8838 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...