Potassium oleate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
O0056-25G
O0056-500G
TCO0056-025GEA
20.35
USD
TCO0056-025G
TCO0056-500G
Potassium oleate ≥98.0% (by titrimetric analysis)
Potassium oleate
CAS Number: 143-18-0
MDL Number: MFCD00064243
Molecular Formula: C18H34O2
Molecular Weight: 320.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Oleic Acid Potassium Salt
Formula:
C₁₈H₃₃KO₂ Melting Pt: 235…240 °C |
MDL Number:
MFCD00064243 CAS Number: 143-18-0 EINECS: 205-590-5 Merck Index: 13,07735 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...