Ethyl-4-nitropyrrole-2-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Nitropyrrole-2-carboxylic Acid Ethyl Ester, Ethyl 4-nitro-1H-pyrrole-2-carboxylate

N0510-5G N0510-1G
TCN0510-005GEA 228.1 USD
TCN0510-005G TCN0510-001G
Ethyl-4-nitropyrrole-2-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-4-nitropyrrole-2-carboxylate

CAS Number: 5930-92-7
MDL Number: MFCD00059927
Molecular Formula: C7H8N2O4
Molecular Weight: 184.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174


Other Product Names / Synonyms: 4-Nitropyrrole-2-carboxylic Acid Ethyl Ester

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00059927
CAS Number: 5930-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71d8b69e399a989ef35d3e42fdf82fc294d4e76c&vid=71d8b69e399a989ef35d3e42fdf82fc294d4e76c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR