Nitric acid 68.0 - 70.0%, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Nitric acid solution
MACR2704-14
MACR2704-69
MK2704-14CS
309.23
USD
MK2704-14
MK2704-69
Nitric acid 68.0 - 70.0%, AR® ACS, Macron Fine Chemicals™
Nitric acid
ACS Reagent meets the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.
Formula:
HNO₃ MW: 63.01 g/mol Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
ACS - Assay (HNO₃) | 68.0 - 70.0 % |
ACS - Color (APHA) | ≤10 |
ACS - Heavy Metals (as Pb) | ≤0.2 ppm |
ACS - Residue after Ignition | ≤5 ppm |
ACS - Sulfate (SO₄) | ≤1 ppm |
Trace Impurities - Arsenic (As) | ≤0.010 ppm |
Chloride (Cl) | ≤0.5 ppm |
Trace Impurities - Iron (Fe) | ≤0.2 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...