2-Methyl-2-butene ≥95.0%

Supplier: TCI America
Danger

Synonyms: Trimethylethylene, Isoamylene, Isopentene

P0067-500ML P0067-25ML P0067-100ML
TCP0067-500MLEA 170.95 USD
TCP0067-500ML TCP0067-25ML TCP0067-100ML
2-Methyl-2-butene ≥95.0%
2-Methyl-2-butene

CAS Number: 513-35-9
MDL Number: MFCD00009276
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 38
Melting point (°C): -134
Flash Point (°C): -45
Specific Gravity (20/20): 0.66


Other Product Names / Synonyms: Amylene, Pentene

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 38 °C
Melting Pt: –134 °C
Density: 0.66 (20 °C)
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009276
CAS Number: 513-35-9
EINECS: 208-156-3
UN: 2460
ADR: 3,II
Merck Index: 12,00649
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=186d47d3ab8b63d9610f4d0b29142b8bc8cf21c7&vid=3a6c9e6ed1cbda67a0588f70b6999e3c5669feb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR