N-(Hydroxymethyl)acrylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0574-25G M0574-500G
TCM0574-025GEA 27.48 USD
TCM0574-025G TCM0574-500G
N-(Hydroxymethyl)acrylamide ≥98.0% (by titrimetric analysis)
N-(Hydroxymethyl)acrylamide

CAS Number: 924-42-5
MDL Number: MFCD00004597
Molecular Formula: C4H7NO2
Molecular Weight: 101.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 277
Melting point (°C): 76
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N-Methylolacrylamide

Formula: C₄H₇NO₂
Boiling Pt: 277 °C
Melting Pt: 76 °C
MDL Number: MFCD00004597
CAS Number: 924-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ca577bf152e04302a46bcab1d4e36cfbc08743&vid=22ca577bf152e04302a46bcab1d4e36cfbc08743&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR