Butyl myristate ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 110-36-1
MDL Number: MFCD00015077
Molecular Formula: C18H36O2
Molecular Weight: 284.48
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 166
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Myristic Acid Butyl Ester
Formula:
C₁₈H₃₆O₂ Boiling Pt: 195 °C Density: 0.86 (20 °C) Flash Pt: 166 °C |
MDL Number:
MFCD00015077 CAS Number: 110-36-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...