Surshield® Safety-Winged Infusion Sets, Terumo®
Supplier: Terumo
SV*S23BL SV*S19BL SV*S23BLS SV*S21BL SV*S25BL SV*S21BLS SV*S25BLS SV-S19BLS
14229-281CS 1644.84 USD
14229-281 14229-278 14229-297 14229-280 14229-284 14229-296 14229-301 76492-664
Surshield® Safety-Winged Infusion Sets, Terumo®
Catheters
Minimize the risk of needlestick injuries by simply pressing the shield over the needle until it locks into place
There is no change in needle insertion required, and the simple, built-in design is easy to use. All needles are 1.9cm (3/4") long. Latex-free.
Información para pedidos: Infusion sets are packaged in packs of 50 with 10 packs per case.