D-(+)-Galactose, white, powder

Supplier: RELIABLE BIOPHARMACEUTICAL LLC.

3441-1KG 3441-12KG 3441-25KG 3441-50KG
77437-884EA 1125.81 USD
77437-884 77437-886 77437-888 77437-890
D-(+)-Galactose, white, powder
D-(+)-Galactose
Formula: C₆H₁₂O₆
Melting Pt: 168...170 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00151230
CAS Number: 59-23-4

Order Now

Specification Test Results

pH 5 - 7 at 180 g/L at 25 °C
Solubility 180 g/L at 20 °C
Source Plant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR