Jackson Safety® G40 Nitrile Coated Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
40226 40227 40225 40229 40228
89003-833CS 837.37 USD
89003-833 89003-835 89003-831 89003-839 89003-837
Jackson Safety® G40 Nitrile Coated Gloves, Kimberly-Clark
Gloves
Designed to provide an improved level of performance and quality, the new G40 nitrile coated glove replaces the G40 PURPLE NITRILE™ glove as the optimal choice for jobs with improved strength, grip, and dexterity.
  • Gloves ensure a better oil and industrial grease grip
  • The washable design maintains the EN388 ratings through at least five wash cycles
  • Improved heat transfer stamp, with ratings visible even after glove use
  • Ecofriendly packaging with no separate insert, no printed bags, and less waste
  • Silicone-free
Typical applications include automotive and aerospace assembly, metal, glass, and wood handling.