Methyl difluoroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22197-14
B22197-06
AAB22197-14EA
222
USD
AAB22197-14
AAB22197-06
AAB22197-03
Methyl difluoroacetate 98%
Methyl difluoroacetate
Purity: 98%
CAS No.: 433-53-4
Molecular Formula: F2CHCO2CH3
Formula Weight: 110.06
Flash Point: 19°(66°F)
Refractive Index: 1.3350
Boiling Point: 86-87°
Beilstein Reference: 1747534
UN No.: 2924
MDL No.: MFCD00039276
Formula:
C₃H₄F₂O₂ MW: 110.06 g/mol Boiling Pt: 86…87 °C Density: 1.272 g/cm³ (20 °C) Flash Pt: 19 °C (66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039276 CAS Number: 433-53-4 EINECS: 207-089-7 UN: 2924 ADR: 3,III |
Specification Test Results
Boiling Point | 86–87°C |
Flash Point: | 19°C |
Refractive Index: | 1.3514 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...