Jackson Safety™ G60 PURPLE NITRILE® Cut Resistant Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
97434 97431
89011-808CS 568.66 USD
89011-808 89011-804 89011-812 89011-806 89011-810
Jackson Safety™ G60 PURPLE NITRILE® Cut Resistant Gloves, Kimberly-Clark
Gloves
New and improved G60 PURPLE NITRILE® gloves provide two times the cut resistance while maintaining dexterity and comfort.
  • Ambidextrous
  • Ideal for metal fabrication, glass/sharp part handling, and automotive assembly
  • Flexible, nitrile-dotted palm for enhanced grip
  • Dyneema® fabric enhances breathability
More tapered coating on the fingers provide added dexterity.
CPPT: 1150 g.
Certificaciones: ANSI level 3.