Lithium dodecyl sulfate ≥99%, white powder, Ultrapure
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
04800752
04800753
IC800752EA
79.52
USD
IC800752
IC800753
Lithium dodecyl sulfate ≥99%, white powder, Ultrapure
Lithium dodecyl sulphate
Anionic detergent that may be used in place of SDS for electrophoresis in cold conditions.
Soluble in water.
Precaución: Research Use Only (RUO).
Formula:
C₁₂H₂₅LiO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00007467 CAS Number: 2044-56-6 EINECS: 218-058-2 UN: 1325 ADR: 4.1,III |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
OD260 (1% aq soln) | ≤0.1 |
OD280 (1% aq soln) | ≤0.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...