Accucore™ AQ Columns
Supplier: Thermo Scientific
17326-154630 17326-052130 17326-014005 17326-013005 17326-012105 17326-152130 17326-104630 17326-103030 17326-102130 17326-053030
10038-798EA 1554.87 USD
10038-798 10038-800 10038-806 10038-808 10038-810 10039-140 10039-144 10039-146 10039-148 10039-150
Accucore™ AQ Columns
Chromatography Columns
Separate polar compounds with improved speed and resolution using Thermo Scientific™ Accucore aQ columns.
Rugged 2.6 µm solid core particles enable fast, high efficiency separations at low backpressures. The polar endcapped C18 stationary phase provides optimal retention of polar analytes and is fully compatible with 100% aqueous mobile phases. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Información para pedidos: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm ID require the 4/4.6 mm UNIGUARD Holder (10046-744).