Accucore™ PFP Columns
Supplier: Thermo Scientific
17426-153030 17426-154630 17426-152130 17426-104630 17426-103030 17426-102130 17426-053030 17426-052130 17426-014005 17426-012105 17426-013005
10038-768EA 1352.93 USD
10038-768 10038-770 10038-772 10038-774 10038-776 10038-778 10038-782 10038-784 10038-792 10038-794 10038-796
Accucore™ PFP Columns
Chromatography Columns
Improve LC separations of halogenated compounds and non-halogenated polar analytes using Thermo Scientific™ Accucore PFP columns.
Rugged 2.6 µm solid core particles enable fast, high-resolution separations at low backpressures. Fluorine groups in the PFP stationary phase provide enhanced retention and selectivity for positional isomers of halogenated compounds as well as for compounds with polar functional groups. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Información para pedidos: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm ID require the 4/4.6 mm UNIGUARD Holder (10046-744).