Isoquinoline-5-sulfonic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

I0470-25G I0470-250G
TCI0470-025GEA 55.4 USD
TCI0470-025G TCI0470-250G
Isoquinoline-5-sulfonic acid ≥96.0% (by HPLC, titration analysis)
Isoquinoline-5-sulphonic acid

CAS Number: 27655-40-9
MDL Number: MFCD00134089
Molecular Formula: C9H7NO3S
Molecular Weight: 209.22
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 5-Sulfoisoquinoline

Formula: C₉H₇NO₃S
MW: 209.22 g/mol
MDL Number: MFCD00134089
CAS Number: 27655-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a47bbefee73047f6738bbd9a24c7bc44e8e4ae&vid=45a47bbefee73047f6738bbd9a24c7bc44e8e4ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR