Hydrogen fluoride pyridine complex pyridine ~30 %, hydrogen fluoride ~70 %

Danger

Synonyms: Olah′s reagent, Pyridinium polybifluoride, Pyridine hydrofluoride, HF-Pyridine, PPHF, Poly(pyridine fluoride), Pyridinium poly(hydrogen fluoride)

M-Clarity™ quality level = MQ200

184225-100G 184225-25G
220040-215EA 148.62 USD
220040-215 220040-216
Hydrogen fluoride pyridine complex pyridine ~30 %, hydrogen fluoride ~70 %
Hydrogen fluoride pyridine complex
Formula: C₅H₅N·(HF)x
Storage Temperature: Freezer
MDL Number: MFCD00012436
CAS Number: 62778-11-4
UN: 1790
ADR: 8 (6.1),I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR