BRAND® PCR Plates, 384-Well, Rigid Frame, BIO-CERT® PCR QUALITY
Supplier: Brandtech
781538 781539
77447-856PK 303.05 USD
77447-856 77447-858
BRAND® PCR Plates, 384-Well, Rigid Frame, BIO-CERT® PCR QUALITY
Microplates PCR Plates
BRAND® 384-well plates offer efficient PCR and qPCR solutions tailored for automatic high-throughput analyses. Featuring a strong polycarbonate frame and thin-walled wells for rapid temperature transfer.
  • Compatible with all standard thermal cyclers and fully automation ready
  • Select from semi-skirted or fully skirted plate options
  • Distinguish workflow steps easily with a range of color choices
  • White plates for qPCR, optimized to reflect fluorescence signals
BRAND® rigid frame PCR plates feature a robust polycarbonate frame, ensuring safe handling in both semi-automated and automated pipetting systems while providing optimal evaporation protection.
The rigid frame design also facilitates the use of ultra-thin-walled wells made from 100% virgin medical-grade polypropylene, ensuring rapid temperature transfer from the cycler to the sample, consequently reducing cycle times. The 384-well rigid frame plates are manufactured in controlled cleanroom environments.
Certificaciones: BIO-CERT® PCR quality level, made from virgin, raw polypropylene, free of human DNA, RNase, DNase, pyrogens, and PCR-inhibitors.