Amidinothiourea ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Guanylthiourea

G0234-25G G0234-500G G0234-100G
TCG0234-025GEA 33 USD
TCG0234-025G TCG0234-500G TCG0234-100G
Amidinothiourea ≥98.0% (by HPLC, titration analysis)
Amidinothiourea

CAS Number: 2114-02-5
MDL Number: MFCD00014472
Molecular Formula: C2H6N4S
Molecular Weight: 118.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₂H₆N₄S
MW: 118.158 g/mol
MDL Number: MFCD00014472
CAS Number: 2114-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a541d836a85ead950fc5f2e72634c7718975eaef&vid=a541d836a85ead950fc5f2e72634c7718975eaef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR