Guanidinium hydrochloride ≥99%, white crystalline powder, Ultrapure
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Guanidine Hydrochloride is a protein denaturant and thus having an important role in molecular weight determinations.
Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine HCl is used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties. It is an inhibitor of RNase. Highly concentrated (6 - 8 M) Guanidine HCl solutions are used to denature native globular proteins. It apparently disrupts hydrogen bonds which hold the protein in its unique structure. However, there also is evidence suggesting that guanidine hydrocholoride may disrupt hydrophobic interactions by promoting the solubility of hydrophobic residues in aqueous solutions.
- Ultra Pure Grade
- Suitable for molecular biology applications
- White Crystalline Powder
- pKa: 13.6 (Lit.)
- Store at Room Temperature (15-30 °C). Store Desiccated.
Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine HCl is used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties. It is an inhibitor of RNase. Highly concentrated (6 - 8 M) Guanidine HCl solutions are used to denature native globular proteins. It apparently disrupts hydrogen bonds which hold the protein in its unique structure. However, there also is evidence suggesting that guanidine hydrocholoride may disrupt hydrophobic interactions by promoting the solubility of hydrophobic residues in aqueous solutions.
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
pH (6.0 M aq soln) | 4-6 |
OD260 (6.0M aq soln) | ≤0.03 |
Loss on Drying | ≤1.0% |
Solubility (6.0 M aq soln) | Clear, Colorless Solution |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...