Guanidinium hydrochloride ≥99%, crystalline powder, Ultrapure
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Applications
Guanidine hydrochloride can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Also used in the isolation of RNA. It is a strong chaotropic agent useful for the denaturation and subsequent refolding of protein, it can solubilize insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Also used in the isolation of RNA.
Product Description
The crystal structure of Guanidine hydrochloride consists of a network of guanidinium cations and chloride anions linked by N–H•••Cl hydrogen bonds,it is a strong chaotropic agent.Guanidine HCl may agglomerate upon storage. It may appear as a free-flowing crystalline powder, a freeflowing powder with solid material dispersed throughout, or a solid. The quality of the product does not appear to be affected and solutions prepared from the free-flowing and lumpy guanidine HCl appear identical
Grade: Ultra Pure
Purity: >99%
Keywords: Guanidium chloride, chaotropic agent
Key Applications: Chaeotropic agent
Product Type: Biochemicals
Biochemical Category: Chaotropic Agents
Density: 1.345 g/cm³ at 20 °C (Lit.)
Melting Point: 180-190 °C
UV/Visible Absorbance: OD260nm (6.0 M aq soln) <0.03
Presentation: White Crystalline Powder
pH: 4-6 (6.0 M aq soln)
Solubility: Soluble in water,Clear, Colorless Solution (6 M - clear, colorless solution).
Storage & Handling: Room Temperature, desiccate
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...