STANSOLV® Knit-Lined Nitrile Gloves, Mapa Professionnel
Supplier: MAPA
34381048 34381049 34381040 34381041
32920-270CS 1258.1 USD
32920-270 32920-272 32920-274 32920-276
STANSOLV® Knit-Lined Nitrile Gloves, Mapa Professionnel
Gloves
STANSOLV® AK-22 gloves provide extreme liquid-tightness in a knit-lined nitrile glove.
  • The lightweight nitrile shell provides excellent comfort and dexterity
  • Medium weight
  • Pinked cuff
Unique, patented separate shell-and-liner construction prevents leak paths caused by liner fibers penetrating the nitrile shell. The soft, two-way-stretch interlock cotton liner absorbs perspiration and helps insulate the glove and guarantees long-lasting protection from permeation by toxic compounds. Unique, embossed Z-pattern grip channels fluids off the working surface to ensure a secure grip even with hard-to-grasp materials. Ideal for chemical processing, construction, maintenance metal fabrication, petrochemical processing, and seafood processing. Color: green.
Certificaciones: USDA-accepted for food and pharmaceutical applications per CFR 21 170-199. CE marked.
Información para pedidos: Packaged one dozen pair per poly bag.