Urea ≥99%, white prills, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

04821519 04821527 04821530 04821528
IC821519EA 147.17 USD
IC821519 IC821527 IC821530 76180-662
Urea ≥99%, white prills, Ultrapure
Urea

Urea is the principal end product of nitrogen metabolism in most mammals, formed by the enzymatic reactions of the Kreb's cycle.
Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels. Urea is used in cell or tissue culture media to increase the osmolality. Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.
Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses. When performing enzymatic protein digests it is important to remove urea first. Even though some enzymes will tolerate small amounts of urea, the elevated temperature used for most reactions will lead to carbamylation during the course of the digest. The urea can be removed prior to digestion by fast reversed phase chromatography, spin columns, or dialysis.
Dissolve urea in deionized water to the desired concentration.For every 10 ml of solution, add 1 g of Amberlite® IRA-910.Stir for one hour at room temperature


  • Ultra Pure Grade
  • Partition Coefficient: log Pow: -2,59 - -1,59
  • One gram dissolves in 1 mL water, 10 mL 95% ethanol, 1 mL boiling 95% ethanol, 20 mL absolute ethanol, 6 mL methanol, 2 mL glycerol; Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
  • Storage temperature: Room Temperature


Denaturation, Protein Electrophoresis, SDS-PAGE


Urea is the principal end product of nitrogen metabolism in most mammals, formed by the enzymatic reactions of the Kreb's cycle which is a mild agent usually used in the solubilization and denaturation of proteins. It is DNAse RNAse free and useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. Urea in solution is in equilibrium with ammonium cyanate ,form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. It has an OD260(7M sq solution) and OD280 as 0.0215 and 0.0049 respectively.


Urea is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels. When performing enzymatic protein digests it is important to remove urea first. Even though some enzymes will tolerate small amounts of urea, the elevated temperature used for most reactions will lead to carbamylation during the course of the digest. It can be removed prior to digest by fast reversed phase chromatography, spin columns, or dialysis. It is used in cell or tissue culture media to increase the osmolality and also used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic. Urea is typically used at a concentration of 8 M for protein denaturation or solubilization. A final concentration of 5 M urea is commonly used in molecular biology for sequencing gels. To prevent carbamylation, do not heat urea containing buffers above 37°C.Ultra pure Urea is used in an experiment to study the role of Vimentin protein as a marker of liver tumorigenicity (Yasushi Nakamura,2009 ,Actin and Vimentin proteins with N-terminal deletion detected in tumor bearing rat livers induced by intraportal-vein injection of Ha-ras transfected rat liver cells ,National Institute Of Health Public Access Author Manuscript,124(11),2512 -2519).

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 135 °C (dec.)
Melting Pt: 130…140 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
OD260 (7M aq soln) ≤0.05
OD280 (7M aq soln) ≤0.05
pH (7M aq soln) 8.5 ± 1.0
Conductivity ≤100 uMhos
Copper ≤1ppm
Lead ≤1ppm
Iron ≤1ppm
Cyanate None Detected
RNAse Negative
DNAse Negative
Protease Negative

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR