Ethylene glycol monoallyl ether ≥98%

Supplier: TCI America
Danger

Synonyms: 2-(Allyloxy)ethanol, 2-Allyloxyethanol, 2-(2-Propenyloxy)ethanol, Allyl glycol

E0326-25ML E0326-500ML
TCE0326-025MLEA 26 USD
TCE0326-025ML TCE0326-500ML
Ethylene glycol monoallyl ether ≥98%
Ethylene glycol monoallyl ether

CAS Number: 111-45-5
MDL Number: MFCD00020607
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 60
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Allyl Glycol, 2-Allyloxyethanol

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 161 °C
Density: 0.96 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00020607
CAS Number: 111-45-5
UN: 3271
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=043a4eb8133e8fde6b0855e04507fab7ef84b9b2&vid=043a4eb8133e8fde6b0855e04507fab7ef84b9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR