4-Ethylbenzaldehyde ≥98.0%

Supplier: TCI America
Warning

Synonyms: p-Ethylbenzaldehyde

E0325-25ML E0325-500ML
TCE0325-025MLEA 25.59 USD
TCE0325-025ML TCE0325-500ML
4-Ethylbenzaldehyde ≥98.0%
4-Ethylbenzaldehyde

CAS Number: 4748-78-1
MDL Number: MFCD00006956
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 92
Specific Gravity (20/20): 1.00

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 221 °C
Density: 1.00 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00006956
CAS Number: 4748-78-1
EINECS: 225-268-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4c143a2bc08e4f9eddf4acdbee6ec050f22841&vid=3b6403becf1672ae3dcc332ba79517d2bca7adb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR