Ethanol ≥95%, KOPTEC, meets analytical specification of BP, Ph. Eur., USP (190 Proof) (not denatured)

Supplier: Decon Labs
Danger

Synonyms: Ethyl alcohol

V1155M V1105 V1105TP V1101 V1105SG V1105HC V1155 V1101TP V1155TP V1101G V1101GTP V1105HCTP V1105SGTP V1155MTP V1105MTP V1105M
89125-162CS 194.78 USD
89125-162 71001-626 89125-166 89125-178 89125-182 89125-164 71001-630 71002-510 89125-168 89125-180 89125-184 71001-868 71002-424 71002-428 89234-854 89234-856 89234-858 89501-406 89501-408
Ethanol ≥95%, KOPTEC, meets analytical specification of BP, Ph. Eur., USP (190 Proof) (not denatured)
Ethanol

190 proof (95%) pure (undenatured) ethanol. Meets USP and multi-compendial specifications. Used for research, healthcare, industrial, and production applications as a cleaner, solvent, and reagent.


  • Water 5%
  • Ethyl alcohol: 95%
  • Denaturant: none

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3
Melting Pt: -117
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Order Now

Specification Test Results

Assay 190 proof 94.9-96.0 (% v/v) Ethanol
Water 4.0-5.1 (% v/v)
Specific Gravity @15.56C 0.8120-0.8160 (USP)
Specific Gravity @15.0C 0.8140-0.8160 (JP)
Relative Density @ 20C 0.8051-0.8124 (BP/EP)
Color Passes USP test for color
Clarity Passes USP Test for Clarity
Acidity Max. 30 ppm
Alkalinity Passes USP Test
Non Volatile Residue Max. 25 ppm
Sum of Acetal & Acetaldehyde compounds Max.10 ppm
Benzene Max.2 ppm
Methanol Max.100 ppm
UV Absorbance @ 240 nm Max. 0.40 AU (USP/BP/EP/JP)
UV Absorbance @ 250 nm Max. 0.30 AU (USP/BP/EP/JP)
UV Absorbance @ 260 nm Max. 0.30 AU (USP/BP/EP/JP)
UV Absorbance @ 270 nm Max. 0.10 AU (USP/BP/EP/JP)
UV Absorbance @ 235-340 nm (Smooth curve) Pass (USP/BP/EP/JP)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR