(3S)-(-)-3-(Dimethylamino)pyrrolidine ≥99.0%

Supplier: TCI America

Synonyms: (S)-3-(Dimethylamino)pyrrolidine, (S)-(-)-3-(Dimethylamino)pyrrolidine

D2193-5G D2193-1G
TCD2193-5GEA 220 USD
TCD2193-5G TCD2193-001G
(3S)-(-)-3-(Dimethylamino)pyrrolidine ≥99.0%
(3S)-(-)-3-(Dimethylamino)pyrrolidine

CAS Number: 132883-44-4
MDL Number: MFCD00191348
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 51
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: -14.5 deg (C=10, EtOH)

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 164 °C
Density: 0.90 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00191348
CAS Number: 132883-44-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4629da32578be27430478552bd0ae51d11f44a3a&vid=4629da32578be27430478552bd0ae51d11f44a3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR