4,5-Dicyanoimidazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile
D2026-25G
D2026-250G
TCD2026-025GEA
101.71
USD
TCD2026-025G
TCD2026-250G
4,5-Dicyanoimidazole ≥98.0% (by HPLC, total nitrogen)
4,5-Dicyanoimidazole
CAS Number: 1122-28-7
MDL Number: MFCD00005194
Molecular Formula: C5H2N4
Molecular Weight: 118.10
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 175
Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis, Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 4,5-Imidazoledicarbonitrile
Formula:
C₅H₂N₄ MW: 118.1 g/mol Melting Pt: 175 °C Density: 1.458 g/cm³ (20 °C) |
MDL Number:
MFCD00005194 CAS Number: 1122-28-7 EINECS: 214-344-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...