1-Docosanol ≥98.0%

Supplier: TCI America

Synonyms: Behenyl alcohol

D0964-500G D0964-25G
TCD0964-500GEA 271.56 USD
TCD0964-500G TCD0964-025G
1-Docosanol ≥98.0%
1-Docosanol

CAS Number: 661-19-8
MDL Number: MFCD00002939
Molecular Formula: C22H46O
Molecular Weight: 326.61
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Flash Point (°C): 198


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols, Antiviral Ingredients
Other Product Names / Synonyms: Behenyl Alcohol

Formula: H₃C(CH₂)₂₀CH₂OH
MW: 326.61 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 72 °C
Density: ~ 0.81 g/cm³ (20 °C)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00002939
CAS Number: 661-19-8
EINECS: 211-546-6
Merck Index: 13,03433
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0053a26b4552ee0296ddc578bca332aafc2df981&vid=ed8c9873a14a45ac1a4c276f654a89051d9dd72f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR