N,N-Dimethyl-1,3-diaminopropane ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Dimethylaminopropylamine, N,N-Dimethyltrimethylenediamine, N,N-Dimethyl-1,3-propanediamine

D0790-25ML D0790-500ML
TCD0790-025MLEA 20.39 USD
TCD0790-025ML TCD0790-500ML
N,N-Dimethyl-1,3-diaminopropane ≥99.0% (by GC, titration analysis)
N,N-Dimethyl-1,3-diaminopropane

CAS Number: 109-55-7
MDL Number: MFCD00008216
Molecular Formula: C5H14N2
Molecular Weight: 102.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 38
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 3-(Dimethylamino)propylamine, N,N-Dimethyltrimethylenediamine

Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 136 °C
Density: 0.82 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00008216
CAS Number: 109-55-7
EINECS: 203-680-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9c50dcd9f622113d8e3fce323b3e150d72c2b9a&vid=f9c50dcd9f622113d8e3fce323b3e150d72c2b9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR